4-(Trifluormethoxy)thiophenol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Trifluoromethoxy)benzenethiol, 4-(Trifluoromethoxy)benzene-1-thiol

PC7439V-1G PC7439V-5G PC7439V-25G
APOSPC7439V-1GEA 45.1 EUR
APOSPC7439V-1G APOSPC7439V-5G APOSPC7439V-25G
4-(Trifluormethoxy)thiophenol 95%
4-(Trifluormethoxy)thiophenol
Formel: C₇H₅F₃OS
Molecular Weight: 194.18 g/mol
Siedepunkt: 64…66 °C/15mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00052312
CAS-Nummer: 169685-29-4
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR