3-(Trifluormethoxy)phenylacetonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Trifluormethoxy)benzylcyanid
PC7440G-1G
APOSPC7440G-1GEA
21.9
EUR
APOSPC7440G-1G
3-(Trifluormethoxy)phenylacetonitril 98%
3-(Trifluormethoxy)phenylacetonitril
Formel:
C₉H₆F₃NO Molecular Weight: 201,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061272 CAS-Nummer: 108307-56-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...