6-Fluoro-2-methyl-1H-indole 98%

Lieferant: Apollo Scientific

PC7443-1G
APOSPC7443-1GEA 132 EUR
APOSPC7443-1G
6-Fluoro-2-methyl-1H-indole 98%
6-Fluoro-2-methyl-1H-indole
Formel: C₉H₈FN
Molecular Weight: 149.165 g/mol
Schmelzpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027714
CAS-Nummer: 40311-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR