3'-(Trifluormethyl)acetanilid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(3-(Trifluormethyl)phenyl)acetamid, Ethansäure(3-(trifluormethyl)phenyl)amid, 3-Acetamidobenzotrifluorid

PC7450-100G
APOSPC7450-100GEA 292 EUR
APOSPC7450-100G
3'-(Trifluormethyl)acetanilid 98%
3'-(Trifluormethyl)acetanilid
Formel: C₉H₈F₃NO
Molecular Weight: 203,16 g/mol
Schmelzpunkt: 104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000383
CAS-Nummer: 351-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR