2-Brom-5-(trifluormethyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

PC7451-1G
APOSPC7451-1GEA 21.5 EUR
APOSPC7451-1G
2-Brom-5-(trifluormethyl)phenylboronsäure 96%
2-Brom-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BBrF₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09258742
CAS-Nummer: 957034-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR