5-Bromo-4-carboxy-2-fluorobenzeneboronic acid 96%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-4-carboxy-2-fluorophenylboronic acid, 4-Borono-2-bromo-5-fluorobenzoic acid

PC7452-1G
APOSPC7452-1GEA 270 EUR
APOSPC7452-1G
5-Bromo-4-carboxy-2-fluorobenzeneboronic acid 96%
5-Bromo-4-carboxy-2-fluorobenzeneboronic acid
Formel: C₇H₅BBrFO₄
Molecular Weight: 262,826 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09475847
CAS-Nummer: 957034-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR