4'-(Trifluormethyl)acetanilid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-(Trifluormethyl)phenyl)acetamid, Ethansäure(4-(trifluormethyl)phenyl)amid, 4-Acetamidobenzotrifluorid
PC7460-5G
APOSPC7460-5GEA
21.9
EUR
APOSPC7460-5G
4'-(Trifluormethyl)acetanilid 98%
4'-(Trifluormethyl)acetanilid
Formel:
C₉H₈F₃NO Molecular Weight: 203,16 g/mol Schmelzpunkt: 150…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013562 CAS-Nummer: 349-97-3 EINECS: 206-495-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...