Ethyl-1-[2-nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate

Lieferant: Apollo Scientific

PC7461-1G
APOSPC7461-1GEA 257 EUR
APOSPC7461-1G
Ethyl-1-[2-nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate
Ethyl-1-[2-nitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate
Formel: C₁₅H₁₇F₃N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01029145
CAS-Nummer: 610259-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR