1-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine hydrochloride

Lieferant: Apollo Scientific

PC7464-1G
APOSPC7464-1GEA 184 EUR
APOSPC7464-1G
1-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine hydrochloride
1-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine hydrochloride
Formel: C₁₂H₁₄F₃N₃O₂·ClH
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09038316
CAS-Nummer: 1185299-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR