Ethyl-2-(trifluormethyl)imidazo[1,2-a]pyridin-3-carboxylat

Lieferant: Apollo Scientific

PC7469-1G
APOSPC7469-1GEA 329 EUR
APOSPC7469-1G
Ethyl-2-(trifluormethyl)imidazo[1,2-a]pyridin-3-carboxylat
Ethyl-2-(trifluormethyl)imidazo[1,2-a]pyridin-3-carboxylat
Formel: C₁₁H₉F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06496237
CAS-Nummer: 108438-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR