1-[(4-Fluoro-5-methyl-2-methylsulphonyl)phenyl]piperidine-4-carboxamide

Lieferant: Apollo Scientific

PC7474-1G
APOSPC7474-1GEA 213 EUR
APOSPC7474-1G
1-[(4-Fluoro-5-methyl-2-methylsulphonyl)phenyl]piperidine-4-carboxamide
1-[(4-Fluoro-5-methyl-2-methylsulphonyl)phenyl]piperidine-4-carboxamide
Formel: C₁₄H₁₉FN₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09264539
CAS-Nummer: 1000339-66-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR