2-Fluoro-5-methylsulphonyl-3-nitrotoluene

Lieferant: Apollo Scientific

PC7476-1G PC7476-5G
APOSPC7476-1GEA 0 EUR
APOSPC7476-1G APOSPC7476-5G
2-Fluoro-5-methylsulphonyl-3-nitrotoluene
2-Fluoro-5-methylsulphonyl-3-nitrotoluene
Formel: C₈H₈FNO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD09264540
CAS-Nummer: 1000339-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR