3-Chlor-4-(methylsulfonyl)benzotrifluorid

Lieferant: Apollo Scientific

PC7481-1G
APOSPC7481-1GEA 263 EUR
APOSPC7481-1G
3-Chlor-4-(methylsulfonyl)benzotrifluorid
3-Chlor-4-(methylsulfonyl)benzotrifluorid
Formel: C₈H₆ClF₃O₂S
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09264544
CAS-Nummer: 1000339-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR