3-[(4-Fluoro-2-methylsulphonyl)phenoxy]benzoic acid

Lieferant: Apollo Scientific

PC7484-1G
APOSPC7484-1GEA 184 EUR
APOSPC7484-1G
3-[(4-Fluoro-2-methylsulphonyl)phenoxy]benzoic acid
3-[(4-Fluoro-2-methylsulphonyl)phenoxy]benzoic acid
Formel: C₁₄H₁₁FO₅S
Schmelzpunkt: 167…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09264548
CAS-Nummer: 1000339-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR