Ethyl-2-(morpholin-4-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

PC7485-1G
APOSPC7485-1GEA 155 EUR
APOSPC7485-1G
Ethyl-2-(morpholin-4-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Ethyl-2-(morpholin-4-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Formel: C₁₁H₁₃F₃N₂O₃S
Molecular Weight: 310,3 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01315302
CAS-Nummer: 338392-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR