2-Bromo-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-4-(trifluoromethyl)thiazole-5-carboxylic acid

PC7487-1G
APOSPC7487-1GEA 300 EUR
APOSPC7487-1G
2-Bromo-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid
2-Bromo-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid
Formel: C₅HBrF₃NO₂S
Molecular Weight: 276,04 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09264552
CAS-Nummer: 162651-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR