2-Methyl-4-trifluoromethyl-1,3-thiazole-5-carboxamide

Lieferant: Apollo Scientific

PC7489-1G
APOSPC7489-1GEA 268 EUR
APOSPC7489-1G
2-Methyl-4-trifluoromethyl-1,3-thiazole-5-carboxamide
2-Methyl-4-trifluoromethyl-1,3-thiazole-5-carboxamide
Formel: C₆H₅F₃N₂OS
Molecular Weight: 210,18 g/mol
Schmelzpunkt: 42…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202348
CAS-Nummer: 339026-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR