1-[(4-Ethylsulphonyl-2-fluoro)phenyl]-3-methylpiperazine

Lieferant: Apollo Scientific

PC7497-1G
APOSPC7497-1GEA 216 EUR
APOSPC7497-1G
1-[(4-Ethylsulphonyl-2-fluoro)phenyl]-3-methylpiperazine
1-[(4-Ethylsulphonyl-2-fluoro)phenyl]-3-methylpiperazine
Formel: C₁₃H₁₉FN₂O₂S
Molecular Weight: 286,37 g/mol
Schmelzpunkt: 155…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09264562
CAS-Nummer: 1000339-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR