2-(Trifluormethyl)benzamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Trifluormethyl)benzoesäureamid
PC7505-100G
PC7505-25G
PC7505-500G
APOSPC7505-100GEA
94.4
EUR
APOSPC7505-100G
APOSPC7505-25G
APOSPC7505-500G
2-(Trifluormethyl)benzamid 98%
2-(Trifluormethyl)benzamid
Formel:
C₈H₆F₃NO Schmelzpunkt: 160…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014798 CAS-Nummer: 360-64-5 EINECS: 206-637-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...