3-(Trifluormethyl)benzamid 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Trifluormethyl)benzoesäureamid

PC7506-1G
APOSPC7506-1GEA 14.7 EUR
APOSPC7506-1G
3-(Trifluormethyl)benzamid 98%
3-(Trifluormethyl)benzamid
Formel: C₈H₆F₃NO
Molecular Weight: 189,14 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014802
CAS-Nummer: 1801-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR