4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC7521-1G
PC7521-5G
PC7521-10G
APOSPC7521-1GEA
0
EUR
APOSPC7521-1G
APOSPC7521-5G
APOSPC7521-10G
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide 98%
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide
Formel:
C₉H₈BrF₃O₂S Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16606069 CAS-Nummer: 934557-65-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...