3,5-Dichloro-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}aniline

Lieferant: Apollo Scientific

Synonyme: 2-(4-Amino-2,6-dichlorophenoxy)-3-(trifluoromethyl)pyridine

PC7527-1G
APOSPC7527-1GEA 183 EUR
APOSPC7527-1G
3,5-Dichloro-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}aniline
3,5-Dichloro-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}aniline
Formel: C₁₂H₇Cl₂F₃N₂O
Molecular Weight: 323,098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109520
CAS-Nummer: 1227955-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR