5-Fluor-4-chinazolon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoroquinazolin-4(1H)-one, 5-Fluoro-4-hydroxyquinazoline

PC7536-1G
APOSPC7536-1GEA 29 EUR
APOSPC7536-1G
5-Fluor-4-chinazolon 98%
5-Fluor-4-chinazolon
Formel: C₈H₅FN₂O
Molecular Weight: 164,13 g/mol
Schmelzpunkt: 238…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09475836
CAS-Nummer: 436-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR