3-Bromo-5-(trifluoromethyl)benzaldehyde oxime 98%
Lieferant: Apollo Scientific
PC7537-1G
APOSPC7537-1GEA
241
EUR
APOSPC7537-1G
3-Bromo-5-(trifluoromethyl)benzaldehyde oxime 98%
3-Bromo-5-(trifluoromethyl)benzaldehyde oxime
Formel:
C₈H₅BrF₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998195 CAS-Nummer: 876132-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...