4-Carboxy-2-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Carboxy-2-fluorobenzeneboronic acid
PC7538-5G
APOSPC7538-5GEA
148
EUR
APOSPC7538-5G
4-Carboxy-2-fluorphenylboronsäure 98%
4-Carboxy-2-fluorphenylboronsäure
Formel:
C₇H₆BFO₄ Molecular Weight: 183,93 g/mol Schmelzpunkt: 213…215 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08458473 CAS-Nummer: 851335-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...