3-Chloro-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}aniline

Lieferant: Apollo Scientific

PC7547-1G
APOSPC7547-1GEA 183 EUR
APOSPC7547-1G
3-Chloro-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}aniline
3-Chloro-4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}aniline
Formel: C₁₂H₈ClF₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11109521
CAS-Nummer: 1099669-15-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR