2-Amino-4-trifluoromethyl-1,3-thiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(trifluoromethyl)thiazol-2-amine, 2-Amino-4-trifluoromethyl-1,3-thiazole, 4-(trifluoromethyl)-1,3-thiazol-2-amine, 4-Trifluoromethyl-thiazole-2ylamine, 2-Amino-4-trifluoromethylthiazole, 4-(Trifluoromethyl)thiazol-2-amine

PC7554-250MG
APOSPC7554-250MGEA 21.9 EUR
APOSPC7554-250MG
2-Amino-4-trifluoromethyl-1,3-thiazole
2-Amino-4-trifluoromethyl-1,3-thiazole
Formel: C₄H₃F₃N₂S
Molecular Weight: 168.14 g/mol
Schmelzpunkt: 65…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00832759
CAS-Nummer: 349-49-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR