2-Amino-4-[(3,5-difluoro)phenyl]-1,3-thiazole

Lieferant: Apollo Scientific

PC7556-1G
APOSPC7556-1GEA 213 EUR
APOSPC7556-1G
2-Amino-4-[(3,5-difluoro)phenyl]-1,3-thiazole
2-Amino-4-[(3,5-difluoro)phenyl]-1,3-thiazole
Formel: C₉H₆F₂N₂S
Molecular Weight: 212,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424572
CAS-Nummer: 676348-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR