Ethyl-2-methylsulphonyl-4-trifluoromethyl-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific

PC7557-1G
APOSPC7557-1GEA 184 EUR
APOSPC7557-1G
Ethyl-2-methylsulphonyl-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Ethyl-2-methylsulphonyl-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Formel: C₈H₈F₃NO₄S₂
Molecular Weight: 303,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750912
CAS-Nummer: 1000339-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR