Ethyl-2-(pyrrolidin-1-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Lieferant: Apollo Scientific
PC7558-1G
APOSPC7558-1GEA
184
EUR
APOSPC7558-1G
Ethyl-2-(pyrrolidin-1-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Ethyl-2-(pyrrolidin-1-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Formel:
C₁₁H₁₃F₃N₂O₂S Molecular Weight: 294,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09750913 CAS-Nummer: 1000339-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...