3-[(2-Fluoro-4-methylsulphonyl)phenoxy]benzoic acid

Lieferant: Apollo Scientific

PC7564-1G
APOSPC7564-1GEA 184 EUR
APOSPC7564-1G
3-[(2-Fluoro-4-methylsulphonyl)phenoxy]benzoic acid
3-[(2-Fluoro-4-methylsulphonyl)phenoxy]benzoic acid
Formel: C₁₄H₁₁FO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD09750919
CAS-Nummer: 1000339-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR