2-(Trifluormethyl)benzophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Phenyl(2-(trifluoromethyl)phenyl)methanone
PC7568-100G
PC7568-25G
PC7568-5G
APOSPC7568-100GEA
410
EUR
APOSPC7568-100G
APOSPC7568-25G
APOSPC7568-5G
2-(Trifluormethyl)benzophenon 98%
2-(Trifluormethyl)benzophenon
Formel:
C₁₄H₉F₃O Schmelzpunkt: 58…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000377 CAS-Nummer: 727-99-1 EINECS: 211-972-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...