4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (4-(Cyclopentylcarbamoyl)-3-fluorophenyl)boronic acid, N-Cyclopentyl 4-borono-2-fluorobenzamide, 4-(Cyclopentylcarbamoyl)-3-fluorophenylboronic acid

PC7573-1G
APOSPC7573-1GEA 243 EUR
APOSPC7573-1G
4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid 98%
4-(Cyclopentylcarbamoyl)-3-fluorobenzeneboronic acid
Formel: C₁₂H₁₅BFNO₃
Molecular Weight: 251.062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09475825
CAS-Nummer: 957034-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR