1-(Benzyloxy)-4-brom-2,3-difluorbenzol 98%

Lieferant: Apollo Scientific
Achtung

PC7574-1G PC7574-5G
APOSPC7574-1GEA 40.6 EUR
APOSPC7574-1G APOSPC7574-5G
1-(Benzyloxy)-4-brom-2,3-difluorbenzol 98%
1-(Benzyloxy)-4-brom-2,3-difluorbenzol
Formel: C₁₃H₉BrF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09475823
CAS-Nummer: 941294-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR