Dimethyl-2-[2-nitro-4-(trifluoromethyl)phenyl]malonate 98%

Lieferant: Apollo Scientific

Synonyme: Dimethyl 2-(2-nitro-4-(trifluoromethyl)phenyl)malonate

PC7575-1G
APOSPC7575-1GEA 168 EUR
APOSPC7575-1G
Dimethyl-2-[2-nitro-4-(trifluoromethyl)phenyl]malonate 98%
Dimethyl-2-[2-nitro-4-(trifluoromethyl)phenyl]malonate
Formel: C₁₂H₁₀F₃NO₆
Molecular Weight: 321,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617804
CAS-Nummer: 290825-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR