3-Fluor-5-(2,2,2-trifluorethoxy)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7576-1G
APOSPC7576-1GEA
292
EUR
APOSPC7576-1G
3-Fluor-5-(2,2,2-trifluorethoxy)phenylboronsäure 98%
3-Fluor-5-(2,2,2-trifluorethoxy)phenylboronsäure
Formel:
C₈H₇BF₄O₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD09475814 CAS-Nummer: 850589-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...