4-Fluor-3-(2,2,2-trifluorethoxy)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC7577-1G
PC7577-250MG
APOSPC7577-1GEA
229
EUR
APOSPC7577-1G
APOSPC7577-250MG
4-Fluor-3-(2,2,2-trifluorethoxy)phenylboronsäure 95%
4-Fluor-3-(2,2,2-trifluorethoxy)phenylboronsäure
Formel:
C₈H₇BF₄O₃ Schmelzpunkt: 116…119 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09475813 CAS-Nummer: 957034-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...