4-Bromo-2,6-difluorophenylboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2,6-difluorobenzeneboronic acid

PC7580-1G
APOSPC7580-1GEA 0 EUR
APOSPC7580-1G
4-Bromo-2,6-difluorophenylboronic acid
4-Bromo-2,6-difluorophenylboronic acid
Formel: C₆H₄BBrF₂O₂
Molecular Weight: 236.807 g/mol
Schmelzpunkt: 155…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03095369
CAS-Nummer: 352535-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR