Methyl perfluoro-3,6-dioxadecanoate
Lieferant: Apollo Scientific
|
Achtung
|
PC7594-25G
PC7594-5G
PC7594-100G
APOSPC7594-25GEA
202
EUR
APOSPC7594-25G
APOSPC7594-5G
APOSPC7594-100G
Methyl perfluoro-3,6-dioxadecanoate
Methyl perfluoro-3,6-dioxadecanoate
Formel:
C₉H₃F₁₅O₄ Siedepunkt: 50 °C/12mmHg Dichte: 1,8 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094429 CAS-Nummer: 129301-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...