5-Brom-2-fluorbenzamid

Lieferant: Apollo Scientific
Achtung

PC7597-1G
APOSPC7597-1GEA 151 EUR
APOSPC7597-1G
5-Brom-2-fluorbenzamid
5-Brom-2-fluorbenzamid
Formel: C₇H₅BrFNO
Molecular Weight: 218,02 g/mol
Schmelzpunkt: 138…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094430
CAS-Nummer: 214210-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR