4-Brom-2-fluorbenzamid

Lieferant: Apollo Scientific
Achtung

PC7599-1G PC7599-5G
APOSPC7599-1GEA 33.5 EUR
APOSPC7599-1G APOSPC7599-5G
4-Brom-2-fluorbenzamid
4-Brom-2-fluorbenzamid
Formel: C₇H₅BrFNO
Molecular Weight: 218.03 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094432
CAS-Nummer: 292621-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR