N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea

Lieferant: Apollo Scientific

PC7606-10G PC7606-1G PC7606-5G
APOSPC7606-10GEA 1350 EUR
APOSPC7606-10G APOSPC7606-1G APOSPC7606-5G
N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea
N-(2-Chloroethyl)-N'-[4-(trifluoromethoxy)phenyl]urea
Formel: C₁₀H₁₀ClF₃N₂O₂
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689716
CAS-Nummer: 937601-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR