2,6-Di-tert-butyl-4-methylpyryliumtrifluormethansulfonat 97%

Lieferant: Apollo Scientific

PC7612-5G
APOSPC7612-5GEA 183 EUR
APOSPC7612-5G
2,6-Di-tert-butyl-4-methylpyryliumtrifluormethansulfonat 97%
2,6-Di-tert-butyl-4-methylpyryliumtrifluormethansulfonat
Formel: C₁₅H₂₃F₃O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00229854
CAS-Nummer: 59643-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR