5-(4-Fluorophenyl)-3,4-dihydro-2H-1,4-thiazine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC7635-1G
APOSPC7635-1GEA 203 EUR
APOSPC7635-1G
5-(4-Fluorophenyl)-3,4-dihydro-2H-1,4-thiazine-3-carboxylic acid
5-(4-Fluorophenyl)-3,4-dihydro-2H-1,4-thiazine-3-carboxylic acid
Formel: C₁₁H₁₀FNO₂S
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689744
CAS-Nummer: 1190102-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR