3-(2-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Lieferant: Apollo Scientific
Achtung

PC7637-1G
APOSPC7637-1GEA 140 EUR
APOSPC7637-1G
3-(2-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
3-(2-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Formel: C₁₄H₉FN₂OS
Molecular Weight: 272,3 g/mol
Schmelzpunkt: 274…276 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00994441
CAS-Nummer: 23892-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR