N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Methoxy-N-methyl-3-(trifluoromethyl)benzamide

PC7638-1G
APOSPC7638-1GEA 58 EUR
APOSPC7638-1G
N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide
N-Methoxy-N-methyl-3-(trifluoromethyl)benzenecarboxamide
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689665
CAS-Nummer: 116332-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR