4-(4-Fluorobenzoyl)butyric acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(4-Fluorophenyl)-5-oxovaleric Acid, 5-(4-Fluorophenyl)-5-oxopentanoic acid

PC7656-500G
APOSPC7656-500GEA 157 EUR
APOSPC7656-500G
4-(4-Fluorobenzoyl)butyric acid
4-(4-Fluorobenzoyl)butyric acid
Formel: C₁₁H₁₁FO₃
Molecular Weight: 210.20 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00667222
CAS-Nummer: 149437-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR