3-(Trifluormethyl)zimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC7663-100G
PC7663-25G
APOSPC7663-100GEA
72.5
EUR
APOSPC7663-100G
APOSPC7663-25G
3-(Trifluormethyl)zimtsäure 98%
3-(Trifluormethyl)zimtsäure
Formel:
C₁₀H₇F₃O₂ Schmelzpunkt: 132…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004393 CAS-Nummer: 779-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...