4-(Trifluormethyl)cyclohexanon 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC7668N-1G
PC7668N-5G
PC7668N-100MG
APOSPC7668N-1GEA
94.4
EUR
APOSPC7668N-1G
APOSPC7668N-5G
APOSPC7668N-100MG
4-(Trifluormethyl)cyclohexanon 97%
4-(Trifluoromethyl)cyclohexanone
Formel:
C₇H₉F₃O Siedepunkt: 67…68 °C/16mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00102145 CAS-Nummer: 75091-99-5 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...