(±)-4-(Trifluormethyl)mandelsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(±)-4-(Trifluormethyl)phenylglykolsäure, (±)-2-(4-(Trifluormethyl)phenyl)-2-hydroxyethansäure, (±)-2-(4-(Trifluormethyl)phenyl)-2-hydroxyessigsäure
PC7687-5G
PC7687-1G
PC7687-25G
APOSPC7687-5GEA
94.8
EUR
APOSPC7687-5G
APOSPC7687-1G
APOSPC7687-25G
(±)-4-(Trifluormethyl)mandelsäure 98%
(±)-4-(Trifluormethyl)mandelsäure
Formel:
C₉H₇F₃O₃ Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009724 CAS-Nummer: 395-35-7 EINECS: 206-900-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...